3,4-Dibromothiophene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR15943-500G OR15943-25G OR15943-100G
76568-016EA 147.7 USD
76568-016 76568-012 76568-014
3,4-Dibromothiophene 98%
3,4-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221
Melting Pt: 4.5
Density: 2.188 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR