4-Bromo-2-methoxythiazole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR15560-1G
OR15560-5G
76567-814EA
75.4
USD
76567-814
76567-816
4-Bromo-2-methoxythiazole
4-Bromo-2-methoxythiazole
Formula:
C₄H₄BrNOS Storage Temperature: Refrigerator |
MDL Number:
MFCD07787374 CAS Number: 240816-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...