1H-Pyrazole-1-carboximidamide hydrochloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine

OR14754-25G OR14754-100G
76567-200EA 20.34 USD
76567-200 76567-202
1H-Pyrazole-1-carboximidamide hydrochloride
1H-Pyrazole-1-carboximidamide hydrochloride
Formula: C₄H₇ClN₄
MW: 146.58 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00210087
CAS Number: 4023-02-3
EINECS: 429-520-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR