5-(4-Methylpiperazino)-2-nitroaniline

Supplier: APOLLO SCIENTIFIC

Synonyms: 5-(4-Methylpiperazin-1-yl)-2-nitroaniline

OR13864-5G OR13864-1G
76566-602EA 49.92 USD
76566-602 76566-600
5-(4-Methylpiperazino)-2-nitroaniline
5-(4-Methylpiperazino)-2-nitroaniline
Formula: C₁₁H₁₆N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00545964
CAS Number: 23491-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR