Diethyl malonate 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester

OR1354-500G OR1354-10KG OR1354-2.5KG
76566-388EA 20.32 USD
76566-388 76566-392 76566-390
Diethyl malonate 99%
Diethyl malonate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199
Melting Pt: -50
Density: 1.055 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR