2,2'-Bithiophene 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR1311-100G OR1311-500G OR1311-25G
76566-144EA 58.06 USD
76566-144 76566-146 76566-142
2,2'-Bithiophene 99%
2,2'-Bithiophene
Formula: C₈H₆S₂
Boiling Pt: 260 °C
Melting Pt: 32…33 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005414
CAS Number: 492-97-7
EINECS: 207-767-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR