4-(2-Pyridyl)benzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde

OR12648-1G OR12648-250MG
76565-866EA 23.34 USD
76565-866 76565-864
4-(2-Pyridyl)benzaldehyde
4-(2-Pyridyl)benzaldehyde
Formula: C₁₂H₉NO
Melting Pt: 51…53 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD01863537
CAS Number: 127406-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR