Homophthalic anhydride
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR12643-100G
OR12643-25G
OR12643-5G
76565-862EA
269.94
USD
76565-862
76565-860
76565-858
Homophthalic anhydride
Homophthalic anhydride
Formula:
C₉H₆O₃ MW: 162.14 g/mol Melting Pt: 140…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006894 CAS Number: 703-59-3 EINECS: 211-873-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...