2-Bromoisonicotinonitrile 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR11615-25G
OR11615-5G
OR11615-1G
76565-342EA
101.88
USD
76565-342
76565-340
76565-338
2-Bromoisonicotinonitrile 95%
2-Bromoisonicotinonitrile
Formula:
C₆H₃BrN₂ Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD07367879 CAS Number: 10386-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...