2-Bromoisonicotinonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR11615-25G OR11615-5G OR11615-1G
76565-342EA 101.88 USD
76565-342 76565-340 76565-338
2-Bromoisonicotinonitrile 95%
2-Bromoisonicotinonitrile
Formula: C₆H₃BrN₂
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD07367879
CAS Number: 10386-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR