Methyl 2,3-dimethylquinoxaline-6-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR110390-1G
76564-072EA 200.96 USD
76564-072
Methyl 2,3-dimethylquinoxaline-6-carboxylate
Methyl 2,3-dimethylquinoxaline-6-carboxylate
Formula: C₁₂H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00205437
CAS Number: 32461-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR