2-Butyl-1H-benzimidazole

Supplier: APOLLO SCIENTIFIC
Warning

OR110234-1G
76563-874EA 66.22 USD
76563-874
2-Butyl-1H-benzimidazole
2-Butyl-1H-benzimidazole
Formula: C₁₁H₁₄N₂
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00714456
CAS Number: 5851-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR