4-Iodo-1H-pyrazole 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR10990-25G OR10990-100G OR10990-500G
76563-584EA 20.26 USD
76563-584 76563-586 76563-588
4-Iodo-1H-pyrazole 99%
4-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS Number: 3469-69-0
EINECS: 222-434-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR