4-Iodo-1H-pyrazole 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR10990-25G
OR10990-100G
OR10990-500G
76563-584EA
20.26
USD
76563-584
76563-586
76563-588
4-Iodo-1H-pyrazole 99%
4-Iodo-1H-pyrazole
Formula:
C₃H₃IN₂ Melting Pt: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005244 CAS Number: 3469-69-0 EINECS: 222-434-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...