3-Bromobenzenesulfonamide

Supplier: APOLLO SCIENTIFIC
Warning

OR0112-1G OR0112-5G
76563-362EA 20.77 USD
76563-362 76561-262
3-Bromobenzenesulfonamide
3-Bromobenzenesulphonamide
Formula: C₆H₆BrNO₂S
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00084903
CAS Number: 89599-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR