(S)-(+)-α-Methoxyphenylacetic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR52197-1G OR52197-25G OR52197-5G
76561-036EA 20.26 USD
76561-036 76563-212 76563-010
(S)-(+)-α-Methoxyphenylacetic acid 95%
(S)-(+)-α-Methoxyphenylacetic acid
Formula: C₉H₁₀O₃
Boiling Pt: 164…166 °C/18mmHg
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00064216
CAS Number: 26164-26-1
EINECS: 247-492-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR