5-Methoxy-1H-indole-3-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR10185-25G OR10185-5G
76563-108EA 103.9 USD
76563-108 76560-354
5-Methoxy-1H-indole-3-carbaldehyde
5-Methoxy-1H-indole-3-carbaldehyde
Formula: C10H9NO2
Melting Pt: 179…184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005623
CAS Number: 10601-19-1
EINECS: 234-220-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR