2-Chlorothioanisole

Supplier: APOLLO SCIENTIFIC
Warning

OR0032-100G OR0032-10G OR0032-25G
76561-974EA 109.42 USD
76561-974 76562-828 76560-608
2-Chlorothioanisole
2-Chlorothioanisole
Formula: C₇H₇ClS
Boiling Pt: 239…240 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00040124
CAS Number: 17733-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR