Calcium chloride 77%

Supplier: APOLLO SCIENTIFIC
Warning

IN1385A-100G IN1385A-1KG IN1385A-50G
76561-272EA 20.07 USD
76561-272 76562-796 76562-908
Calcium chloride 77%
Calcium chloride
Formula: CaCl₂
Boiling Pt: 1670 °C
Melting Pt: 772 °C
Density: 2.15 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR