2-Chloro-3-pyridineboronic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR9357-100G OR9357-25G OR9357-5G
76562-706EA 287.26 USD
76562-706 76563-114 76560-502
2-Chloro-3-pyridineboronic acid 96%
2-Chloro-3-pyridineboronic acid
Formula: C₅H₅BClNO₂
Melting Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094997
CAS Number: 381248-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR