1-Chloro-3-(triphenylphosphoranylidene)acetone

Supplier: APOLLO SCIENTIFIC
Warning

OR10637-250MG
76560-724EA 195.7 USD
76560-724 76562-700
1-Chloro-3-(triphenylphosphoranylidene)acetone
1-Chloro-3-(triphenylphosphoranylidene)acetone
Formula: C₂₁H₁₈ClOP
Storage Temperature: Ambient
MDL Number: MFCD04038921
CAS Number: 13605-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR