Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR01979-1G OR01979-250MG
76562-684EA 288.54 USD
76562-684 76559-810
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₈Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09842623
CAS Number: 1000018-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR