4-(4-Fluorophenoxy)benzonitrile

Supplier: APOLLO SCIENTIFIC

PC410059-1G PC410059-5G
76562-516EA 62.14 USD
76562-516 76562-578
4-(4-Fluorophenoxy)benzonitrile
4-(4-Fluorophenoxy)benzonitrile
Formula: C₁₃H₈FNO
MDL Number: MFCD01815090
CAS Number: 215589-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR