2-(Piperidin-2-ylmethyl)-3,3,3-trifluoropropanoic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC4651-250MG
76562-550EA 99.82 USD
76562-550
2-(Piperidin-2-ylmethyl)-3,3,3-trifluoropropanoic acid
2-(Piperidin-2-ylmethyl)-3,3,3-trifluoropropanoic acid
Formula: C₉H₁₄F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD03427205
CAS Number: 480438-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR