5-Bromo-2-methox-6-methylypyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR01795-1G OR01795-5G
76559-768EA 20.77 USD
76559-768 76562-542
5-Bromo-2-methox-6-methylypyridine
5-Bromo-2-methox-6-methylypyridine
Formula: C₇H₈BrNO
Boiling Pt: 212 °C
Density: 1.468 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094943
CAS Number: 126717-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR