4-Bromo-2,5-dimethoxytoluene 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR0132-1G OR0132-250MG
76562-338EA 65.2 USD
76562-338 76559-994
4-Bromo-2,5-dimethoxytoluene 97+%
4-Bromo-2,5-dimethoxytoluene
Formula: C₉H₁₁BrO₂
Boiling Pt: 160 °C/30mmHg
Melting Pt: 81…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00666999
CAS Number: 13321-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR