L(+)-Potassium sodium tartrate tetrahydrate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

IN2941-100G
76562-192EA 90.57 USD
76562-192
L(+)-Potassium sodium tartrate tetrahydrate
L(+)-Potassium sodium tartrate tetrahydrate
Formula: KOOC(CHOH)₂COONa·4H₂O
Melting Pt: 70…80 °C
Density: 1.79 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR