4-Hydroxyphenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

OR10505-25G OR10505-5G OR10505-100G
76562-158EA 33.62 USD
76562-158 76561-704 76562-648
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR