4-Quinolinecarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR0731-5G OR0731-1G OR0731-25G
76559-714EA 81.5 USD
76559-714 76562-134 76561-916
4-Quinolinecarboxylic acid
4-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
Melting Pt: 253…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00006782
CAS Number: 486-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR