4-Amino-3-chloro-5-methylbenzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR017872-250MG
76562-122EA 20.26 USD
76562-122
4-Amino-3-chloro-5-methylbenzoic acid 95%
4-Amino-3-chloro-5-methylbenzoic acid
Formula: C₈H₈ClNO₂
Melting Pt: 282…284 °C
Storage Temperature: Ambient
MDL Number: MFCD01313709
CAS Number: 157069-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR