(S)-(+)-β-Aminophenethyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

OR8743-100G OR8743-25G OR8743-500G
76562-052EA 43.8 USD
76562-052 76561-482 76563-392
(S)-(+)-β-Aminophenethyl alcohol
(S)-(+)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00064404
CAS Number: 20989-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR