4,5-Dicyanoimidazole 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile

OR10446-25G
76559-444EA 67.98 USD
76559-444 76561-890 76562-388
4,5-Dicyanoimidazole 98%
4,5-Dicyanoimidazole
Formula: C₅H₂N₄
MW: 118.1 g/mol
Melting Pt: 168…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00005194
CAS Number: 1122-28-7
EINECS: 214-344-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR