Methyl-4-bromo-3-methylbenzoate

Supplier: APOLLO SCIENTIFIC
Warning

OR0924-1G OR0924-25G OR0924-5G
76561-834EA 20.77 USD
76561-834 76559-840 76561-758
Methyl-4-bromo-3-methylbenzoate
Methyl 4-bromo-3-methylbenzoate
Formula: C₉H₉BrO₂
Melting Pt: 38…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00673014
CAS Number: 148547-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR