Methyl-4-bromo-3-methylbenzoate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR0924-1G
OR0924-25G
OR0924-5G
76561-834EA
20.77
USD
76561-834
76559-840
76561-758
Methyl-4-bromo-3-methylbenzoate
Methyl 4-bromo-3-methylbenzoate
Formula:
C₉H₉BrO₂ Melting Pt: 38…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673014 CAS Number: 148547-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...