Niobium(V) oxide 99.99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Diniobium pentaoxide, Niobium pentoxide

IN2773-100G IN2773-500G
76562-754EA 260.16 USD
76562-754 76561-652
Niobium(V) oxide 99.99%
Niobium (V) oxide
Formula: Nb₂O₅
Melting Pt: 1485 °C
Density: 4.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011128
CAS Number: 1313-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR