5-Chloroindole 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
5-Chloro-1H-indole
OR10199-10G
OR10199-25G
OR10199-5G
76561-372EA
38.72
USD
76561-372
76561-458
76563-252
5-Chloroindole 97%
5-Chloroindole
Formula:
C₈H₆ClN MW: 151.6 g/mol Melting Pt: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005672 CAS Number: 17422-32-1 EINECS: 241-448-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...