(3,4-Dichlorophenyl)acetic acid methyl ester

Supplier: APOLLO SCIENTIFIC
Warning

OR0617-10G
76561-396EA 20.26 USD
76561-396 76562-140 76562-894
(3,4-Dichlorophenyl)acetic acid methyl ester
(3,4-Dichlorophenyl)acetic acid methyl ester
Formula: (C₆H₃Cl₂)CH₂COOCH₃
Melting Pt: 25…27 °C
Density: 1.33 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00800680
CAS Number: 6725-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR