Methyl-2-fluoro-4-nitrobenzoate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Methyl 2-fluoro-4-nitrobenzoate
PC8742-10G
PC8742-1G
PC8742-5G
76561-348EA
33.48
USD
76561-348
76559-986
76559-804
Methyl-2-fluoro-4-nitrobenzoate
Methyl 2-fluoro-4-nitrobenzoate
Formula:
C₈H₆FNO₄ Melting Pt: 73…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD08444028 CAS Number: 392-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...