3-Bromo-N-(2-fluorophenyl)propanamide

Supplier: APOLLO SCIENTIFIC
Warning

PC31961-1G PC31961-250MG
76561-176EA 563.68 USD
76561-176 76562-510
3-Bromo-N-(2-fluorophenyl)propanamide
3-Bromo-N-(2-fluorophenyl)propanamide
Formula: C₉H₉BrFNO
Storage Temperature: Ambient
MDL Number: MFCD00124372
CAS Number: 7661-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR