1-Bromo-3,5-diiodobenzene 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR54592-1G
76561-062EA 75.4 USD
76561-062
1-Bromo-3,5-diiodobenzene 95%
1-Bromo-3,5-diiodobenzene
Formula: C₆H₃BrI₂
Storage Temperature: Ambient
MDL Number: MFCD07778997
CAS Number: 149428-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR