4-(Methylsulfonyl) phenylacetonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR0210-1G OR0210-250MG
76561-548EA 75.4 USD
76561-548 76561-030
4-(Methylsulfonyl) phenylacetonitrile
4-(Methylsulfonyl) phenylacetonitrile
Formula: C₉H₉NO₂S
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00079775
CAS Number: 25025-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR