Calcium fluoride, anhydrous 99.99%

Supplier: APOLLO SCIENTIFIC
Warning

IN1387-5G
76560-932EA 20.26 USD
76560-932
Calcium fluoride, anhydrous 99.99%
Calcium fluoride
Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500
Melting Pt: 1418
Density: 3.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR