4-Bromo-2-fluoro-3-methylbenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC48590-1G PC48590-250MG PC48590-5G
76560-346EA 385.06 USD
76560-346 76560-906 76561-000
4-Bromo-2-fluoro-3-methylbenzonitrile
4-Bromo-2-fluoro-3-methylbenzonitrile
Formula: C₈H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD23707391
CAS Number: 1114546-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR