Pyridazine-4,5-dicarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR1036-500MG OR1036-250MG
76559-864EA 28.2 USD
76559-864 76560-592
Pyridazine-4,5-dicarboxylic acid
Pyridazine-4,5-dicarboxylic acid
Formula: C₆H₄N₂O₄
Melting Pt: >210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06742784
CAS Number: 59648-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR