2,2-Dichlorocyclopropanecarboxamide

Supplier: APOLLO SCIENTIFIC
Warning

OR0348-1G
76560-586EA 69.26 USD
76560-586
2,2-Dichlorocyclopropanecarboxamide
2,2-Dichlorocyclopropanecarboxamide
Formula: C₄H₅Cl₂NO
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00218854
CAS Number: 75885-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR