(±)-α-Methoxyphenylacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR13894-1G OR13894-500MG OR13894-5G
76559-912EA 23.32 USD
76559-912 76560-510 76561-552
(±)-α-Methoxyphenylacetic acid
(±)-α-Methoxyphenylacetic acid
Formula: C₉H₁₀O₃
Boiling Pt: 165 °C/18mmHg
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00064217
CAS Number: 7021-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR