N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide

Supplier: APOLLO SCIENTIFIC
Warning

PC7638-1G
76560-408EA 54.54 USD
76560-408
N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide
N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide
Formula: C₁₀H₁₀F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD08689665
CAS Number: 116332-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR