5-(Phenylethynyl)-2-furoic acid, Technical Grade
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR0707-1G
OR0707-250MG
76560-292EA
131.8
USD
76560-292
76561-406
76561-976
5-(Phenylethynyl)-2-furoic acid, Technical Grade
5-(Phenylethynyl)-2-furoic acid
Formula:
C₁₃H₈O₃ Melting Pt: 212…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00541949 CAS Number: 130423-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...