4-(Benzyloxy)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10362-100G OR10362-25G OR10362-5G
76560-270EA 131.4 USD
76560-270 76562-512 76560-100
4-(Benzyloxy)phenylboronic acid 98%
4-(Benzyloxy)phenylboronic acid
Formula: C₁₃H₁₃BO₃
Melting Pt: 198…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075705
CAS Number: 146631-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR