2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%

Supplier: APOLLO SCIENTIFIC

PC48602-1G
76560-234EA 20.34 USD
76560-234
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine
Formula: C₆H₂ClF₃IN
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD10699394
CAS Number: 205444-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR